ENA118036870 Enamine

3-(1,3-dithiolan-2-yl)aniline

  • MDL number MFCD06255148
  • Cas Number : 50427-03-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 193...195
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-22706

Safety Information

msds https://www.enaminestore.com/catalog/EN300-22706
Availability details for
Quantity
  • Availability
  • Add to Cart