ENA118036873 Enamine

2-[(2-bromobenzoyl)amino]-4-(methylsulfonyl)butanoic acid

  • MDL number MFCD09373402
  • Cas Number : 1032039-60-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23081

Safety Information

msds https://www.enaminestore.com/catalog/EN300-23081
Availability details for
Quantity
  • Availability
  • Add to Cart