ENA118036883 Enamine

5-(chloromethyl)-N-(4-methylphenyl)-1,3,4-thiadiazole-2-carboxamide

  • MDL number MFCD09383891
  • Cas Number : 113940-14-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23638

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23638
Availability details for
Quantity
  • Availability
  • Add to Cart