ENA150998768 Enamine

4-chloro-N-[(4-fluorophenyl)methyl]-N-{[4-(hydrazinecarbonyl)phenyl]methyl}benzene-1-sulfonamide

  • MDL number MFCD09802044
  • Cas Number : 1000930-52-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27236

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27236
Availability details for
Quantity
  • Availability
  • Add to Cart