ENA150998772 Enamine

methyl 4-({N-[(pyridin-3-yl)methyl]4-methoxybenzenesulfonamido}methyl)benzoate

  • MDL number MFCD09802106
  • Cas Number : 1000931-00-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-28941

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28941
Availability details for
Quantity
  • Availability
  • Add to Cart