ENA150998774 Enamine

4-(1H-1,2,4-triazol-1-yl)benzaldehyde oxime

  • MDL number MFCD09802123
  • Cas Number : 920502-11-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-29197

Safety Information

msds https://www.enaminestore.com/catalog/EN300-29197
Availability details for
Quantity
  • Availability
  • Add to Cart