ENA150998877 Enamine

5-(trifluoromethyl)-1-[3-(trifluoromethyl)phenyl]-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD09802052
  • Cas Number : 98534-85-1
  • Molecular Weight : 324.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-27812

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27812
Availability details for
Quantity
  • Availability
  • Add to Cart