ENA150998884 Enamine

1-(4-fluorophenyl)-5-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD03934807
  • Cas Number : 618070-62-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-28259

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28259
Availability details for
Quantity
  • Availability
  • Add to Cart