ENA150998886 Enamine

[2-(4-methoxyphenyl)-1,3-oxazol-4-yl]acetonitrile

  • MDL number MFCD09802058
  • Cas Number : 949980-19-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-28559

Safety Information

msds https://www.enaminestore.com/catalog/EN300-28559
Availability details for
Quantity
  • Availability
  • Add to Cart