ENA150998889 Enamine

5-[(2-bromophenyl)methyl]-1,3-thiazol-2-amine

  • MDL number MFCD08443054
  • Cas Number : 926258-87-9
  • Molecular Weight : 269.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-28582

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28582
Availability details for
Quantity
  • Availability
  • Add to Cart