ENA150998899 Enamine

methyl 2-chloro-5-(diethylsulfamoyl)benzoate

  • MDL number MFCD00814541
  • Cas Number : 345624-82-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-28651

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28651
Availability details for
Quantity
  • Availability
  • Add to Cart