ENA150998921 Enamine

methyl [1,3]dioxolo[4,5-g]cinnoline-3-carboxylate

  • MDL number MFCD09802091
  • Cas Number : 949682-15-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-28886

Safety Information

msds https://www.enaminestore.com/catalog/EN300-28886
Availability details for
Quantity
  • Availability
  • Add to Cart