ENA150998930 Enamine

1-bromo-4-[(4-nitrobenzyl)oxy]benzene

  • MDL number MFCD03999068

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-15516

Safety Information

msds https://www.enaminestore.com/catalog/EN300-15516
Availability details for
Quantity
  • Availability
  • Add to Cart