ENA150998954 Enamine

N-methyl-N-[2-(trifluoromethyl)benzyl]amine

  • MDL number MFCD02689994
  • Cas Number : 296276-41-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-28978

Safety Information

msds https://www.enaminestore.com/catalog/EN300-28978
Availability details for
Quantity
  • Availability
  • Add to Cart