ENA150998958 Enamine

2,5,6-trimethyl-4-(1H-tetrazol-5-yl)pyridazin-3(2H)-one

  • MDL number MFCD09802117
  • Cas Number : 1955530-28-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-28995

Safety Information

msds https://www.enaminestore.com/catalog/EN300-28995
Availability details for
Quantity
  • Availability
  • Add to Cart