ENA150998959 Enamine

N-(1-phenylethyl)urea

  • MDL number MFCD00014792
  • Cas Number : 40589-84-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 121...123
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-28996

Safety Information

msds https://www.enaminestore.com/catalog/EN300-28996
Availability details for
Quantity
  • Availability
  • Add to Cart