ENA150998966 Enamine

1-(cyclopropylcarbonyl)piperidin-4-amine

  • MDL number MFCD07186465
  • Cas Number : 883106-47-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 53...55
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-29007

Safety Information

msds https://www.enaminestore.com/catalog/EN300-29007
Availability details for
Quantity
  • Availability
  • Add to Cart