ENA150998969 Enamine

1-{4'-[(pyrrolidin-1-yl)methyl]-[1,1'-biphenyl]-2-yl}methanamine

  • MDL number MFCD09049744
  • Cas Number : 926187-27-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-29011

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-29011
Availability details for
Quantity
  • Availability
  • Add to Cart