ENA150998974 Enamine

4-fluoro-2-nitro-5-(1H-pyrazol-1-yl)aniline

  • MDL number MFCD09802122
  • Cas Number : 1014020-15-5
  • Molecular Weight : 222.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-29182

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-29182
Availability details for
Quantity
  • Availability
  • Add to Cart