ENA150998980 Enamine

2-nitro-9H-xanthen-9-one

  • MDL number MFCD00691592
  • Cas Number : 20061-59-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-29206

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-29206
Availability details for
Quantity
  • Availability
  • Add to Cart