ENA150998983 Enamine

3,3'-dibromo-5,5'-di-tert-butyl-1,1'-biphenyl-4,4'-diol

  • MDL number MFCD00546819
  • Cas Number : 60803-31-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 116...119
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-18481

Safety Information

msds https://www.enaminestore.com/catalog/EN300-18481
Availability details for
Quantity
  • Availability
  • Add to Cart