ENA150998992 Enamine

N-(2-methoxyethyl)piperidine-4-carboxamide

  • MDL number MFCD06739723
  • Cas Number : 73415-61-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-29223

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-29223
Availability details for
Quantity
  • Availability
  • Add to Cart