ENA150999009 Enamine

2-(4-methylphenyl)-4,5-dihydro-1H-imidazole

  • MDL number MFCD00463854
  • Cas Number : 13623-58-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-29398

Safety Information

msds https://www.enaminestore.com/catalog/EN300-29398
Availability details for
Quantity
  • Availability
  • Add to Cart