ENA150999012 Enamine

N-(4-cyanophenyl)-2-methoxyacetamide

  • MDL number MFCD03576145
  • Cas Number : 104703-39-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-29402

Safety Information

msds https://www.enaminestore.com/catalog/EN300-29402
Availability details for
Quantity
  • Availability
  • Add to Cart