ENA150999014 Enamine

4-(2-chloroacetamido)-3-nitrobenzoic acid

  • MDL number MFCD09802138
  • Cas Number : 949873-89-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-29417

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-29417
Availability details for
Quantity
  • Availability
  • Add to Cart