ENA150999015 Enamine

3-(2-phenylacetamido)propanoic acid

  • MDL number MFCD00458998
  • Cas Number : 55154-47-7
  • Molecular Weight : 207.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-18577

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-18577
Availability details for
Quantity
  • Availability
  • Add to Cart