ENA150999024 Enamine

1-(4-bromophenyl)-5-methyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD04122758
  • Cas Number : 187998-44-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 211...213
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-29442

Safety Information

msds https://www.enaminestore.com/catalog/EN300-29442
Availability details for
Quantity
  • Availability
  • Add to Cart