ENA150999040 Enamine

3-chloro-5,6-dimethylpyridazine-4-carbonitrile

  • MDL number MFCD09802146
  • Cas Number : 93824-72-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 79-80
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-29459

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-29459
Availability details for
Quantity
  • Availability
  • Add to Cart