ENA150999049 Enamine

2-cyano-N-(3-cyanothiophen-2-yl)acetamide

  • MDL number MFCD03946326
  • Cas Number : 73227-83-5
  • Molecular Weight : 191.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-29480

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-29480
Availability details for
Quantity
  • Availability
  • Add to Cart