ENA150999051 Enamine

5-[(2-cyclohexyl-1,3-thiazol-4-yl)methyl]-1,3,4-oxadiazol-2-amine

  • MDL number MFCD03789550
  • Cas Number : 885951-37-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-29487

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-29487
Availability details for
Quantity
  • Availability
  • Add to Cart