ENA150999068 Enamine

3-(3-chloropropanoyl)-1-cyclopentylurea

  • MDL number MFCD09802157
  • Cas Number : 949791-72-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-29506

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-29506
Availability details for
Quantity
  • Availability
  • Add to Cart