ENA150999069 Enamine

(4-acetylphenyl)urea

  • MDL number MFCD00031456
  • Cas Number : 13143-02-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-29507

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-29507
Availability details for
Quantity
  • Availability
  • Add to Cart