ENA150999072 Enamine

4-methylpiperidine-1-sulfonyl chloride

  • MDL number MFCD08445024
  • Cas Number : 41483-70-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-29511

Safety Information

msds https://www.enaminestore.com/catalog/EN300-29511
Availability details for
Quantity
  • Availability
  • Add to Cart