ENA150999075 Enamine

8-(bromomethyl)-5-nitroquinoline

  • MDL number MFCD09046595
  • Cas Number : 89159-94-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-29514

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-29514
Availability details for
Quantity
  • Availability
  • Add to Cart