ENA150999087 Enamine

9-(oxiran-2-ylmethyl)-9H-carbazole

  • MDL number MFCD00218388
  • Cas Number : 52131-82-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-29535

Safety Information

msds https://www.enaminestore.com/catalog/EN300-29535
Availability details for
Quantity
  • Availability
  • Add to Cart