ENA150999097 Enamine

2-(4-nitrophenyl)-1,3,4-oxadiazole

  • MDL number MFCD00491617
  • Cas Number : 4291-13-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 155...157
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-29551

Safety Information

msds https://www.enaminestore.com/catalog/EN300-29551
Availability details for
Quantity
  • Availability
  • Add to Cart