ENA150999103 Enamine

2-(4-methylpiperazine-1-carbonyl)aniline

  • MDL number MFCD03013018
  • Cas Number : 93288-86-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-29564

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-29564
Availability details for
Quantity
  • Availability
  • Add to Cart