ENA150999110 Enamine

3-amino-3-thien-3-ylpropanoic acid

  • MDL number MFCD00152179
  • Cas Number : 94333-62-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-29575

Safety Information

msds https://www.enaminestore.com/catalog/EN300-29575
Availability details for
Quantity
  • Availability
  • Add to Cart