ENA150999117 Enamine

2-(4-methylbenzenesulfonyl)ethanethioamide

  • MDL number MFCD09802170
  • Cas Number : 54334-58-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-29583

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-29583
Availability details for
Quantity
  • Availability
  • Add to Cart