ENA150999118 Enamine

3-(chloromethyl)-5-(2-chlorophenyl)-1,2,4-oxadiazole

  • MDL number MFCD08444943
  • Cas Number : 110704-33-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 83...85
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-29584

Safety Information

msds https://www.enaminestore.com/catalog/EN300-29584
Availability details for
Quantity
  • Availability
  • Add to Cart