ENA150999127 Enamine

6-(1H-pyrazol-1-yl)pyridin-3-amine

  • MDL number MFCD08700232
  • Cas Number : 154095-29-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 97...99
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-29598

Safety Information

msds https://www.enaminestore.com/catalog/EN300-29598
Availability details for
Quantity
  • Availability
  • Add to Cart