ENA150999132 Enamine

2-[3-(pyridin-2-yloxy)phenoxy]acetic acid

  • MDL number MFCD09802175
  • Cas Number : 950028-97-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-29607

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-29607
Availability details for
Quantity
  • Availability
  • Add to Cart