ENA150999133 Enamine

2-[2-oxo-3-(trifluoromethyl)-1,2-dihydropyridin-1-yl]acetic acid

  • MDL number MFCD09802176
  • Cas Number : 950114-48-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-29608

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-29608
Availability details for
Quantity
  • Availability
  • Add to Cart