ENA150999134 Enamine

2-[(1,7-dimethyl-3-oxo-2,3-dihydro-1H-inden-4-yl)oxy]acetic acid

  • MDL number MFCD08445193
  • Cas Number : 923249-44-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-29610

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-29610
Availability details for
Quantity
  • Availability
  • Add to Cart