ENA150999149 Enamine

3-(4-nitro-1H-imidazol-1-yl)propanoic acid

  • MDL number MFCD09802185
  • Cas Number : 145192-61-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-29634

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-29634
Availability details for
Quantity
  • Availability
  • Add to Cart