ENA150999158 Enamine

methyl 4-[(chloroacetyl)amino]butanoate

  • MDL number MFCD09802005
  • Cas Number : 94359-90-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23133

Safety Information

msds https://www.enaminestore.com/catalog/EN300-23133
Availability details for
Quantity
  • Availability
  • Add to Cart