ENA150999159 Enamine

2-methyl-5,6,7,8-tetrahydroquinazolin-6-amine

  • MDL number MFCD08447508
  • Cas Number : 929973-41-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 56...58
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-29654

Safety Information

msds https://www.enaminestore.com/catalog/EN300-29654
Availability details for
Quantity
  • Availability
  • Add to Cart