ENA150999179 Enamine

2-(2,3-dimethoxyphenyl)-1,3-thiazole-4-carboxylic acid

  • MDL number MFCD09802194
  • Cas Number : 949977-50-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-29684

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-29684
Availability details for
Quantity
  • Availability
  • Add to Cart