ENA150999181 Enamine

N-(4-carbamothioylphenyl)acetamide

  • MDL number MFCD09802196
  • Cas Number : 4714-68-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-29687

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-29687
Availability details for
Quantity
  • Availability
  • Add to Cart