ENA150999184 Enamine

5-(chloromethyl)-3-(2-furyl)-1,2,4-oxadiazole

  • MDL number MFCD07636853
  • Cas Number : 501653-22-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 82...84
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-29694

Safety Information

msds https://www.enaminestore.com/catalog/EN300-29694
Availability details for
Quantity
  • Availability
  • Add to Cart